missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoyl Peroxide - Contains 25% water, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
Specifications
Specifications
| Chemical Name or Material | Benzoyl Peroxide |
| CAS | 94-36-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C14 H10 O4 |
| Quantity | 2.5 g |
| Synonym | Benzoyl peroxide hydrous, Peroxide, dibenzoyl, Benzoyl peroxide (8CI), Dibenzoyl peroxide, 617P37, Abcat 40, Acetoxyl, Acne-Aid Cream, Acnegel, Acnezoyl, Akneroxide L, Aksil 5, Aztec BP 50FT, B 75W, BP, BP 50FT, BPO, BPO 50, Basiron, Benbel C, Benox 50, Benox A 80, Benoxyl, Benzac, Benzac W, Benzagel, Benzagel 10, Benzaknen, Benzashave, Benzoic peroxyanhydride, Benzoperoxide, Benzoyl superoxide, Brevoxyl, C 1A, C 1A (peroxide), CH 50, CH 50L, Cadet BPO, Cadet BPO 78W, Cadox 40E, Cadox B, Cadox B 40E, Cadox B 40ES, Cadox B 50P, Cadox B 75W, Cadox B-CH 50, Chaloxyd BP 50FT, Clear By Design, Debroxide, Degament H, Desanden, Desquam E, Desquam X, Diphenylglyoxal peroxide, Dry and Clear, Epi-Clear, Fivenox B 50G, G 20, Interox BP 50P1, Interox GZ-S, Link-Cup DBP, Loroxide, Lucidol, Lucidol (peroxide), Lucidol 40E, Lucidol 50P, Lucidol 70, Lucidol 75, Lucidol 75FP, Lucidol 75W, Lucidol 78, Lucidol 98, Lucidol B 50, Lucidol BT 50, Lucidol BW 50T, Lucidol CH 50, Lucidol CH 50L, Lucidol CH 50X, Lucidol G 20, Lucidol KL 50, Lucidol S 50, Lucidol W 40, Luperco AA, Luperco ANS, Luperco AST, Luperox 78, Luperox 98, Luperox A 40FP-EZ9, Luperox A 70S, Luperox A 75, Luperox A 75FP, Luperox A 98, Luperox ACP 35, Luperox AFR 40, Luperox ANS 50G, Luperox ATC 50, Luprox A 75C, NSC 671, NSC 675, Nericur, Niper BMT, Nyper B, Nyper BD, Nyper BMT, Nyper BMT 40, Nyper BMT 40SV, Nyper BMT-K, Nyper BO, Nyper BO-Y, Nyper BS, Nyper BW, Nyper F, Nyper FF, Nyper FF-K, Nyper NS, Oxy-5, Oxy-L, Oxycare 42, Oxylite, PEROX-B 75, PEROX-B 95, PHisoAc BP, Panoxyl, Perkadox 20S, Abcat 40, Acetoxyl, Acne-Aid Cream, Acnegel, Acnezoyl, Akneroxide L, Aksil 5, Aztec BP 50FT, B 75W, BP 50FT, BPO, BPO 50, Basiron, Benbel C, Benox 50, Benox A 80, Benoxyl, Benzac, Benzac W, Benzagel, Benzagel 10, Benzaknen, Benzashave, Benzoperoxide, Benzoyl Superoxide, Brevoxyl, C 1A, C 1A (peroxide), CH 50, CH 50L, Cadet BPO, Cadet BPO 78W, Cadox 40E, Cadox B, Cadox B 40E, Cadox B 40ES, Cadox B 50P, Cadox B 75W, Cadox B-CH 50, Chaloxyd BP 50FT, Clear By Design, Debroxide, Degament H, Desanden, Desquam E, Desquam X, Dibenzoyl peroxide, Diphenylglyoxal peroxide, Dry and Clear, Epi-Clear, Fivenox B 50G, G 20, Interox BP 50P1, Interox GZ-S, Link-Cup DBP, Loroxide, Lucidol, Lucidol (peroxide), Lucidol 40E, Lucidol 50P, Lucidol 70, Lucidol 75, Lucidol 75FP, Lucidol 75W, Lucidol 78, Lucidol 98Perkadox 33, Perkadox BT 50, Perkadox BTW 50, Perkadox CH 50, Perkadox CH 50L, Perkadox CH 50X, Perkadox L 40RPS, Perkadox L 50S, Perkadox L 50S-PS, Perkadox L-DFG, Perkadox L-W 40, Perkadox L-W 40TCP, Perkadox L-W 75, Perkadox PM 50S, Peroxan BP 50PF1, Peroxan BP 50SE, Peroxan BP Powder 50W, Peroxyderm, Persa-Gel, Persadox, Preoxydex, Sanoxit, Sanperox BPO, SikaFast 5200B, Solugel, Solugel (peroxide), Superox 46-750, Superox 744, TC 1, TC 1 (peroxide), Theraderm, Triaz, Vanoxide, Varox ANS, W 75, W 75 (peroxide), Xerac BP 10, Xerac BP 5, Abcat 40, Acetoxyl, Acne-Aid Cream, Acnegel, Acnezoyl, Akneroxide L, Aksil 5, Aztec BP 50FT, B 75W, BP 50FT, BPO, BPO 50, Basiron, Benbel C, Benox 50, Benox A 80, Benoxyl, Benzac, Benzac W, Benzagel, Benzagel 10, Benzaknen, Benzashave, Benzoperoxide, Benzoyl Superoxide, Brevoxyl, C 1A, C 1A (peroxide), CH 50, CH 50L, Cadet BPO, Cadet BPO 78W, Cadox 40E, Cadox B, Cadox B 40E, Cadox B 40ES, Cadox B 50P, Cadox B 75W, Cadox B-CH 50, Chaloxyd BP 50FT, Clear By Design, Debroxide, Degament H, Desanden, Desquam E, Desquam X, Dibenzoyl peroxide, Diphenylglyoxal peroxide, Dry and Clear, Epi-Clear, Fivenox B 50G, G 20, Interox BP 50P1, Interox GZ-S, Link-Cup DBP, Loroxide, Lucidol, Lucidol (peroxide), Lucidol 40E, Lucidol 50P, Lucidol 70, Lucidol 75, Lucidol 75FP, Lucidol 75W, Lucidol 78, Lucidol 98 |
| InChI Formula | InChI=1S/C14H10O4/c15-13(11-7-3-1-4-8-11)17-18-14(16)12-9-5-2-6-10-12/h1-10H |
| SMILES | O=C(OOC(=O)c1ccccc1)c2ccccc2 |
| IUPAC Name | benzoyl benzenecarboperoxoate |
| Molecular Weight (g/mol) | 242.23 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction