missing translation for 'onlineSavingsMsg'
Learn More
Learn More
15-Acetyl Deoxynivalenol (~90%), TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | 15-Acetyl-deoxynivalenol |
| CAS | 88337-96-6 |
| Molecular Formula | C17 H22 O7 |
| Quantity | 2.5 mg |
| Synonym | (3α,7α)-15-(Acetyloxy)-12,13-epoxy-3,7-dihydroxytrichothec-9-en-8-one, Spiro[2,5-methano-1-benzoxepin-10,2'-oxirane], trichothec-9-en-8-one deriv., 15-Acetyldeoxynivalenol, 15-Acetylvomitoxin, 15-O-Acetyl-4-deoxynivalenol, Deoxynivalenol 15-acetate |
| InChI Formula | InChI=1S/C17H22O7/c1-8-4-11-16(6-22-9(2)18,13(21)12(8)20)15(3)5-10(19)14(24-11)17(15)7-23-17/h4,10-11,13-14,19,21H,5-7H2,1-3H3/t10-,11-,13-,14-,15-,16-,17+/m1/s1 |
| SMILES | CC(=O)OC[C@]12[C@H](O)C(=O)C(=C[C@H]1O[C@@H]3[C@H](O)C[C@@]2(C)[C@]34CO4)C |
| IUPAC Name | [(1R,2R,3S,7R,9R,10R,12S)-3,10-dihydroxy-1,5-dimethyl-4-oxospiro[8-oxatricyclo[7.2.1.02,7]dodec-5-ene-12,2'-oxirane]-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 338.35 |
| Formula Weight | 338.1366 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction