missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5Alpha-Androstane-3Alpha,17Beta-diol-d3, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | 5alpha-Androstane-3alpha,17beta-diol-d3 |
| CAS | 79037-33-5 |
| Molecular Formula | C19H29D3O2 |
| Quantity | 2.5 mg |
| Synonym | (3α,5α,17β)-Androstane-3,17-diol-d3, 3α,17β-Dihydroxy-5α-androstane-d3, 3α,5α-Tetrahydrotestosterone-d3, Adiol-d3, Androstane-3α,17β-diol-d3, Etiocholane-3α,17β-diol-d3, Hombreol-d3, NSC 9899-d3, 5α-Androstane-3α,17β- diol-16,16,17-d3 |
| InChI Formula | InChI=1S/C19H32O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12-17,20-21H,3-11H2,1-2H3/t12-,13+,14?,15-,16-,17-,18-,19-/m0/s1/i6D2,17D |
| SMILES | O[C@@]1([2H])C([2H])([2H])C[C@@]2([H])C3CC[C@@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])CC[C@@]21C |
| IUPAC Name | (3R,5S,9S,10S,13S,14S,17S)-16,16,17-trideuterio-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15-dodecahydro-1H-cyclopenta[a]phenanthrene-3,17-diol |
| Molecular Weight (g/mol) | 295.48 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction