missing translation for 'onlineSavingsMsg'
Learn More

2,6,8-Trimethylnon-5-en-4-one (>80%), TRC

Product Code. 30456782
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456782 100 mg 100mg
30449641 250 mg 250mg
30451939 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456782 Supplier Toronto Research Chemicals Supplier No. TRCT796456100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19044-66-7
Molecular Formula C12H22O
Molecular Weight (g/mol) 182.3
Synonym 2,6,8-Trimethyl-5-nonen-4-one
IUPAC Name (E)-2,6,8-trimethylnon-5-en-4-one
SMILES CC(C)C\C(=C\C(=O)CC(C)C)\C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H22O/c1-9(2)6-11(5)8-12(13)7-10(3)4/h8-10H,6-7H2,1-5H3/b11-8+
Formula Weight 182.1671
Percent Purity >95%
Chemical Name or Material 2,6,8-Trimethylnon-5-en-4-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.