missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diaminopurine-2’-deoxyriboside, TRC

Product Code. 30137651
Change view
Click to view available options
Quantity:
25 mg
250 mg
5 g
Unit Size:
250mg
25mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137651 25 mg 25mg
30007169 5 g 5g
30008128 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137651 Supplier Toronto Research Chemicals Supplier No. TRCD41673525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4546-70-7
Molecular Formula C10H14N6O3
Molecular Weight (g/mol) 266.26
Synonym Adenosine, 2-amino-2'-deoxy-, 9H-Purine, 2,6-diamino-9-(2-deoxy-β-D-erythro-pentofuranosyl)- (7CI,8CI), 2-Amino-2'-deoxyadenosine, 2,6-Diamino-9-(2-deoxy-β-D-erythro-pentofuranosyl)purine, 2,6-Diamino-9-(2'-deoxy-β-D-ribofuranosyl)purine, 2,6-Diaminopurine deoxyriboside, 2,6-Diaminopurine-2'-deoxyriboside, 2-Amino-2'-desoxyadenosine, NSC 104303, 9-(2-Deoxy-β-D-erythro-pentofuranosyl)-9H-purin-2,6-diamine
SMILES Nc1nc(N)c2ncn([C@H]3C[C@H](O)[C@@H](CO)O3)c2n1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H14N6O3/c11-8-7-9(15-10(12)14-8)16(3-13-7)6-1-4(18)5(2-17)19-6/h3-6,17-18H,1-2H2,(H4,11,12,14,15)/t4-,5+,6+/m0/s1
Formula Weight 266.1127
Chemical Name or Material 2,6-Diaminopurine-2'-deoxyriboside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.