missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diaminopurine, 98%

Product Code. 11497557
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497557 1 g 1g
11407567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11497557 Supplier Thermo Scientific Alfa Aesar Supplier No. A15649.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1904-98-9 | C5H6N6 | 150.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1904-98-9
Molecular Formula C5H6N6
Molecular Weight (g/mol) 150.145
MDL Number MFCD00213668
InChI Key MSSXOMSJDRHRMC-UHFFFAOYSA-N
Synonym 2,6-diaminopurine, 9h-purine-2,6-diamine, 2-aminoadenine, 1h-purine-2,6-diamine, 2,6-diamino-9h-purine, purine, 2,6-diamino, purine-2,6-diyldiamine, unii-49p95bau4z, ccris 923, purine-2,6-diamine
PubChem CID 30976
ChEBI CHEBI:40235
IUPAC Name 7H-purine-2,6-diamine
SMILES C1=NC2=C(N1)C(=NC(=N2)N)N

Specifications

Melting Point ∼302°C
Quantity 1 g
Beilstein 9624
Merck Index 14,2985
Formula Weight 150.14
Percent Purity 98%
Chemical Name or Material 2,6-Diaminopurine
compliance-icons
Product Identifier
  • 2,6-Diaminopurine
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1H-Purine-2,6-diamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.