missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diaminopurine, 98%

Product Code. 10544731 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10544731 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10544731 Supplier Thermo Scientific Acros Supplier No. 295270050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1904-98-9 | C5H6N6 | 150.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1904-98-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 302°C
Color White to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C5H6N6
MDL Number MFCD00071537
Quantity 5 g
Merck Index 15, 2987
Synonym 2,6-diaminopurine, 9h-purine-2,6-diamine, 2-aminoadenine, 1h-purine-2,6-diamine, 2,6-diamino-9h-purine, purine, 2,6-diamino, purine-2,6-diyldiamine, unii-49p95bau4z, ccris 923, purine-2,6-diamine
Solubility Information Solubility in water: 2.38g/L water (20°C)
InChI Key MSSXOMSJDRHRMC-UHFFFAOYSA-N
SMILES C1=NC2=C(N1)C(=NC(=N2)N)N
IUPAC Name 7H-purine-2,6-diamine
Molecular Weight (g/mol) 150.14
PubChem CID 30976
ChEBI CHEBI:40235
Formula Weight 150.14
Percent Purity 98%
Physical Form Crystalline Powder or Granules
Chemical Name or Material 2, 6-Diaminopurine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Diaminopurine
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.