missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Diaminotoluene-alpha,alpha,alpha-d3, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2,6-Diaminotoluene-d3 |
| CAS | 362049-58-9 |
| Molecular Formula | C7 2H3 H7 N2 |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C7H10N2/c1-5-6(8)3-2-4-7(5)9/h2-4 H,8-9H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])c1c(N)cccc1N |
| IUPAC Name | 2-(trideuteriomethyl)benzene-1,3-diamine |
| Molecular Weight (g/mol) | 125.19 |
| Formula Weight | 125.1032 g/mol |
| Percent Purity | 97 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction