missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-4-fluorophenol, TRC

Product Code. 30369644
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30369644

Brand: Toronto Research Chemicals TRCD425723100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,6-Dibromo-4-fluorophenol
CAS 344-20-7
Molecular Formula C6 H3 Br2 F O
Quantity 100 mg
Synonym 2,6-Dibromo-4-fluorophenol (ACI), 4-Fluoro-2,6-dibromophenol, NSC 190640
InChI Formula InChI=1S/C6H3Br2FO/c7-4-1-3(9)2-5(8)6(4)10/h1-2,10H
SMILES Oc1c(Br)cc(F)cc1Br
IUPAC Name 2,6-dibromo-4-fluoro-phenol
Molecular Weight (g/mol) 269.894
Formula Weight 267.8535
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.