missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromo-4-methylphenol, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 2,6-Dibromo-4-methylphenol |
| CAS | 2432-14-6 |
| Molecular Formula | C7H6Br2O |
| Quantity | 500 mg |
| Synonym | 2,6-Dibromo-(6CI,7CI,8CI)-p-cresol, 2,6-Dibromo-4-cresol, 2,6-Dibromo-p-cresol, Dibromocresol, NSC 76596 |
| InChI Formula | InChI=1S/C7H6Br2O/c1-4-2-5(8)7(10)6(9)3-4/h2-3,10H,1H3 |
| SMILES | Cc1cc(Br)c(O)c(Br)c1 |
| IUPAC Name | 2,6-dibromo-4-methylphenol |
| Molecular Weight (g/mol) | 265.93 |
| Formula Weight | 263.8785 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction