missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromonaphthalene, 99%

Product Code. 15264764 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15264764 1 g Glass bottle 1g
15274764 5 g Glass bottle 5g
15284764 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15264764 Supplier Thermo Scientific Acros Supplier No. 449340010
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13720-06-4 | C10H6Br2 | 285.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13720-06-4
Molecular Formula C10H6Br2
Molecular Weight (g/mol) 285.97
MDL Number MFCD01026462
InChI Key PJZDEYKZSZWFPX-UHFFFAOYSA-N
Synonym naphthalene, 2,6-dibromo, 2,6-dibromo-naphthalene, 2,6-dibromo naphthalene, 2.6-dibromonaphthalene, 2,6-?dibromonaphthalene, acmc-1c7c1, ksc490o3h, 2,6-dibromonaphthalene
PubChem CID 640591
IUPAC Name 2,6-dibromonaphthalene
SMILES BrC1=CC2=CC=C(Br)C=C2C=C1

Specifications

Melting Point 162°C to 167°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 285.97
Percent Purity 99%
Chemical Name or Material 2, 6-Dibromonaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.