missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromoquinone-4-chloroimide, TRC

Product Code. 30138693
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138693 1 g 1g
30132812 5 g 5g
17923105 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138693 Supplier Toronto Research Chemicals Supplier No. TRCD4255601G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 537-45-1
Molecular Formula C6H2Br2ClNO
Molecular Weight (g/mol) 299.35
Synonym 2,6-Dibromo-4-(chloroimino)-2,5-cyclohexadien-1-one, 2,6-Dibromo-N-chloro-p-benzoquinone Imine, 2,6-Dibromo-1,4-benzoquinone-4-chlorimide, 2,6-Dibromo-N-chloro-p-benzoquinoneimine, 2,6-Dibromoquinone chlorimide, 2,6-Dibromoquinone chloroimide, 2,6-Dibromoquinone chloroimine, 2,6-Dibromoquinone-4-chloroimide, 2,6-Dibromoquinone-4-chloroimine, 4-(Chloroimino)-2,6-dibromo-2,5-cyclohexadien-1-one, BQC reagent, N-Chloro-2,6-dibromoquinoneimine, NSC 528
IUPAC Name 2,6-dibromo-4-chloroimino-cyclohexa-2,5-dien-1-one
SMILES O=C1C(Br)=CC(=NCl)C=C1Br

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H2Br2ClNO/c7-4-1-3(10-9)2-5(8)6(4)11/h1-2H
Formula Weight 296.8192
Chemical Name or Material 2,6-Dibromoquinone-4-chloroimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.