missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-nitropyridine, 90%, Tech.

Product Code. 10753381 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10753381 10 g Glass bottle 10g
10114350 50 g Glass bottle 50g
10289443 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10753381 Supplier Thermo Scientific Acros Supplier No. 186000100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 16013-85-7 | C5H2Cl2N2O2 | 192.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16013-85-7
Molecular Formula C5H2Cl2N2O2
Molecular Weight (g/mol) 192.99
MDL Number MFCD00006234
InChI Key SHCWQWRTKPNTEM-UHFFFAOYSA-N
Synonym 2,6-dichloro-3-nitro-pyridine, pyridine, 2,6-dichloro-3-nitro, 3-nitro-2,6-dichloropyridine, 2,6-dichloro-3-nitro pyridine, zlchem 223, pubchem9234, acmc-209dko, 2,6-dichloronitropyridine, dichloronitropyridine, ksc174q3n
PubChem CID 85239
IUPAC Name 2,6-dichloro-3-nitropyridine
SMILES C1=CC(=NC(=C1[N+](=O)[O-])Cl)Cl

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 58°C to 68°C
Color Colorless to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula Cl2(C5H2N)NO2
Quantity 10 g
Formula Weight 192.99
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material 2, 6-Dichloro-3-nitropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dichloro-3-nitropyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.