missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-nitropyridine, 97%

Product Code. 11414787
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414787 10 g 10g
11424787 50 g 50g
11434787 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414787 Supplier Thermo Scientific Alfa Aesar Supplier No. B21255.09
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 16013-85-7 | C5H2Cl2N2O2 | 192.983 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16013-85-7
Molecular Formula C5H2Cl2N2O2
Molecular Weight (g/mol) 192.983
MDL Number MFCD00006234
InChI Key SHCWQWRTKPNTEM-UHFFFAOYSA-N
Synonym 2,6-dichloro-3-nitro-pyridine, pyridine, 2,6-dichloro-3-nitro, 3-nitro-2,6-dichloropyridine, 2,6-dichloro-3-nitro pyridine, zlchem 223, pubchem9234, acmc-209dko, 2,6-dichloronitropyridine, dichloronitropyridine, ksc174q3n
PubChem CID 85239
IUPAC Name 2,6-dichloro-3-nitropyridine
SMILES C1=CC(=NC(=C1[N+](=O)[O-])Cl)Cl

Specifications

Melting Point 62°C to 65°C
Flash Point >110°C (230°F)
Quantity 10 g
Formula Weight 192.99
Percent Purity 97%
Chemical Name or Material 2,6-Dichloro-3-nitropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.