missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals

Product Code. 11334386
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334386 1 g 1g
11344386 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334386 Supplier Thermo Scientific Alfa Aesar Supplier No. L20003.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 55304-75-1 | Molecular Formula: C6H2Cl2F3N | Molecular Weight: 215.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55304-75-1
Molecular Formula C6H2Cl2F3N
Molecular Weight (g/mol) 215.98
MDL Number MFCD00042245
InChI Key UPWAAFFFSGQECJ-UHFFFAOYSA-N
Synonym 2,6-dichloro-3-trifluoromethyl pyridine, pyridine, 2,6-dichloro-3-trifluoromethyl, 2,6-dichloro-3-trifluoromethyl-pyridine, 2,6-dichloro-3-trifluoromethyl pyridine, puriss, pubchem9238, acmc-20a2ib, 2,6-dichloro-5-trifluoromethylpyridine
PubChem CID 1810337
IUPAC Name 2,6-dichloro-3-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC=C(Cl)N=C1Cl

Specifications

Boiling Point 194°C to 196°C
Refractive Index 1.4845
Quantity 1 g
UN Number UN2810
Beilstein 1570287
Formula Weight 215.99
Percent Purity 98%
Chemical Name or Material 2,6-Dichloro-3-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.