missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(chloromethyl)pyridine, Thermo Scientific™

Product Code. p-4510106
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10497622

Brand: Thermo Scientific Maybridge MO00840CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 101990-72-1
Molecular Formula C6H4Cl3N
Molecular Weight (g/mol) 196.455
InChI Key UYBVOPLURVHJOX-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-chloromethyl pyridine, pyridine, 2,6-dichloro-4-chloromethyl, acmc-20dwvl, 2,6-dichloro-isonicotinyl chloride, 2,6-dichloro-4-chloromethyl-pyridine, 4-chloromethyl-2,6-dichloropyridine, pyridine, 2,6-dichloro-4-chloromethyl-9ci, pyridine,2,6-dichloro-4-chloromethyl
PubChem CID 2794626
IUPAC Name 2,6-dichloro-4-(chloromethyl)pyridine
SMILES C1=C(C=C(N=C1Cl)Cl)CCl

Specifications

Assay Percent Range 95%
Quantity 250 mg
Formula Weight 196.46
Percent Purity 95%
Chemical Name or Material 2, 6-Dichloro-4-(chloromethyl)pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.