missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-methoxyaniline, TRC

Product Code. 30384426
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384426 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384426 Supplier Toronto Research Chemicals Supplier No. TRCD435090250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6480-66-6
Molecular Formula C7 H7 Cl2 N O
Quantity 250 mg
InChI Formula InChI=1S/C7H7Cl2NO/c1-11-4-2-5(8)7(10)6(9)3-4/h2-3H,10H2,1H3
SMILES COc1cc(Cl)c(N)c(Cl)c1
IUPAC Name 2,6-dichloro-4-methoxyaniline
Molecular Weight (g/mol) 192.04
Formula Weight 190.9905
Chemical Name or Material 2,6-Dichloro-4-methoxyaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.