missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-nitroaniline (Dichloran), TRC

Product Code. 30137730
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137730 100 g 100g
17958439 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137730 Supplier Toronto Research Chemicals Supplier No. TRCD431750100G
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-30-9
Molecular Formula C6 H4 Cl2 N2 O2
Molecular Weight (g/mol) 207.01
Synonym Aniline, 2,6-dichloro-4-nitro- (6CI,8CI), 2,6-Dichloro-4-nitrobenzenamine, 1-Amino-2,6-dichloro-4-nitrobenzene, 2,6-Dichloro-4-nitroaniline, 2,6-Dichloro-p-nitroaniline, 4-Nitro-2,6-dichloroaniline, AL 50, Allisan, Batran, Bortran, Botran, Botran 75B, Botran 75W, CNA, DCNA, DCNA (fungicide), Dichloran, Dichloran (amine fungicide), Dicloran, Ditranil, NSC 218, Resisan, U-2069
IUPAC Name 2,6-dichloro-4-nitroaniline
SMILES Nc1c(Cl)cc(cc1Cl)[N+](=O)[O-]

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H4Cl2N2O2/c7-4-1-3(10(11)12)2-5(8)6(4)9/h1-2H,9H2
Formula Weight 205.965
Chemical Name or Material Dicloran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.