missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-pyridinemethanol, TRC

Product Code. 30385154
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385154 5 g 5g
30375701 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385154 Supplier Toronto Research Chemicals Supplier No. TRCD4364355G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101990-69-6
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.02
Synonym (2,6-Dichloropyridin-4-yl)methanol, 2,6-Dichloro-4-(hydroxymethyl)pyridine, 2,6-Dichloro-4-pyridinemethanol, 2,6-Dichloropyrid-4-ylcarbinol
IUPAC Name (2,6-dichloropyridin-4-yl)methanol
SMILES OCC1=CC(Cl)=NC(Cl)=C1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H5Cl2NO/c7-5-1-4(3-10)2-6(8)9-5/h1-2,10H,3H2
Formula Weight 176.97
Chemical Name or Material 2,6-Dichloro-4-pyridinemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.