missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(trifluoromethyl)pyridine, 95%, Thermo Scientific™

Product Code. 15279906 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15279906 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15279906 Supplier Thermo Scientific Acros Supplier No. 454050010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Specifications

CAS 39890-98-7
Boiling Point 70°C to 75°C (25.0 mmHg)
Flash Point 85°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Molecular Formula C6H2Cl2F3N
MDL Number MFCD00042246
Quantity 1 g
Synonym 2,6-dichloro-4-trifluoromethyl pyridine, 2,6-dichloro-4-trifluoromethyl-pyridine, 4-trifluoromethyl-2,6-dichloropyridine, 2,6-dichloro-4-trifluoromethylpyridine, acmc-1air6, abbypharma ap-30-1899, pyridine, 2,6-dichloro-4-trifluoromethyl
InChI Key KVNQWVYYVLCZKK-UHFFFAOYSA-N
SMILES FC(F)(F)C1=CC(Cl)=NC(Cl)=C1
IUPAC Name 2,6-dichloro-4-(trifluoromethyl)pyridine
Molecular Weight (g/mol) 215.98
PubChem CID 520968
Formula Weight 215.99
Percent Purity 95%
Chemical Name or Material 2, 6-Dichloro-4-(trifluoromethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dichloro-4-(trifluoromethyl)pyridine, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.