missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-7-methylpurine, 98%, Thermo Scientific™

Product Code. 15505976 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15505976 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15505976 Supplier Thermo Scientific Acros Supplier No. 457440010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2273-93-0
Melting Point 192.0°C to 195.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C6H4Cl2N4
Quantity 1 g
Synonym 2,6-dichloro-7-methyl-7h-purine, 7h-purine, 2,6-dichloro-7-methyl, purine, 2,6-dichloro-7-methyl, 2,6-dichlor-7-methyl-7h-purin, zlchem 60, pubchem9788, 5-26-11-00258 beilstein handbook reference, purine,6-dichloro-7-methyl, 2,6-dichloro-7-methyl-purin, 2,6-dichloro-7-methyl-purine
InChI Key HVMUWHZAZGTMJK-UHFFFAOYSA-N
SMILES CN1C=NC2=C1C(=NC(=N2)Cl)Cl
IUPAC Name 2,6-dichloro-7-methylpurine
Molecular Weight (g/mol) 203.03
PubChem CID 75281
Formula Weight 203.03
Percent Purity 98%
Chemical Name or Material 2,6-Dichloro-7-methylpurine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.