missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroaniline, 98%

Product Code. p-104318 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10237063 25 g Glass bottle 25g
10676102 100 g Glass bottle 100g
10223950 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10237063 Supplier Thermo Scientific Acros Supplier No. 113100250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 608-31-1 | C6H5Cl2N | 162.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-31-1
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.01
MDL Number MFCD00007675
InChI Key JDMFXJULNGEPOI-UHFFFAOYSA-N
Synonym benzenamine, 2,6-dichloro, 2,6-dichlorobenzenamine, 2,6 dichloraniline, 2,6-dichloro aniline, 2,6-dichloranilin, 2,6-dichloro-phenylamine, 2,6-dichlorophenylamine, 2,5-dichlorobenzoic acid pestanal, clonidine impurity c, 2,6-dichloro-aniline
PubChem CID 11846
ChEBI CHEBI:46630
IUPAC Name 2,6-dichloroaniline
SMILES NC1=C(Cl)C=CC=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 35.0°C to 39.0°C
Color White to Brown
Boiling Point 228.0°C
Flash Point 118°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3NH2
Quantity 25 g
Beilstein 12, 626
Solubility Information Solubility in water: 1.6g/L (20°C); 2.6g/L (40°C)
Formula Weight 162.02
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 2, 6-Dichloroaniline, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,6-Dichloroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.