missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number 11474060
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
50-30-6
C7H4Cl2O2
191.01
MFCD00002418
MRUDNSFOFOQZDA-UHFFFAOYSA-N
benzoic acid, 2,6-dichloro, 2,6-dichloro-benzoic acid, unii-634ri764qt, 2,6 dichlorobenzoic acid, 2,6-dichlorobenzoicacid, 2,6-dichloro-benzoicacid, ccris 8610, pubchem20028, acmc-1auop, 2.6-dichlorobenzoic acid
5758
CHEBI:48623
2,6-dichlorobenzoic acid
OC(=O)C1=C(Cl)C=CC=C1Cl
This item is not returnable. View return policy
50-30-6
C7H4Cl2O2
191.01
MFCD00002418
MRUDNSFOFOQZDA-UHFFFAOYSA-N
benzoic acid, 2,6-dichloro, 2,6-dichloro-benzoic acid, unii-634ri764qt, 2,6 dichlorobenzoic acid, 2,6-dichlorobenzoicacid, 2,6-dichloro-benzoicacid, ccris 8610, pubchem20028, acmc-1auop, 2.6-dichlorobenzoic acid
5758
CHEBI:48623
2,6-dichlorobenzoic acid
OC(=O)C1=C(Cl)C=CC=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 141°C to 145°C
Quantity 5 g
Beilstein 973858
Formula Weight 191.01
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'