missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzonitrile, TRC

Product Code. 30387808
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
This item is not returnable. View return policy

Product Code. 30387808

Brand: Toronto Research Chemicals TRCD431945100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichlobenil
CAS 1194-65-6
Molecular Formula C7 H3 Cl2 N
Quantity 100 g
Synonym 2,6-Dichlorobenzonitrile, 2,6-Dichlorocyanobenzene, Casoron, Casoron 133, Casoron 4G, Casoron CS, Casoron G, DBN, DBN (pesticide), Dichlobenil, H 133, NSC 521490, Nia 5996, Niagara 5996, Surfassol
InChI Formula InChI=1S/C7H3Cl2N/c8-6-2-1-3-7(9)5(6)4-10/h1-3H
SMILES Clc1cccc(Cl)c1C#N
IUPAC Name 2,6-dichlorobenzonitrile
Molecular Weight (g/mol) 172.01
Formula Weight 170.9643
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.