missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzylideneacetone, 98%, Thermo Scientific™

Product Code. 11367555
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367555 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11367555 Supplier Thermo Scientific Alfa Aesar Supplier No. L05706.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 81559-89-9
Boiling Point 135°C to 136°C (2 mmHg)
Molecular Formula C10H8Cl2O
Refractive Index 1.592
MDL Number MFCD00051609
Quantity 5 g
Synonym 2,6-dichlorobenzylideneacetone, 3e-4-2,6-dichlorophenyl but-3-en-2-one, 4-2,6-dichlorophenyl but-3-en-2-one, benzene, 1,3-dichloro-2-2-propynyloxy methyl, e-4-2,6-dichlorophenyl-3-buten-2-one, e-4-2,6-dichlorophenyl but-3-en-2-one, e-4-2,6-dichloro-phenyl-but-3-en-2-one
InChI Key QUUBBGLIJLKARO-AATRIKPKSA-N
SMILES CC(=O)C=CC1=C(C=CC=C1Cl)Cl
IUPAC Name (E)-4-(2,6-dichlorophenyl)but-3-en-2-one
Molecular Weight (g/mol) 215.073
PubChem CID 5708785
Formula Weight 215.08
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorobenzylideneacetone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.