missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroindophenol, sodium salt hydrate, 90+%

Product Code. 13565020 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13565020 5 g Glass bottle 5g
13505050 25 g Glass bottle 25g
13515050 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13565020 Supplier Thermo Scientific Acros Supplier No. 450700050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1266615-56-8 | C12H6Cl2NNaO2 | 290.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1266615-56-8
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key CVSUAFOWIXUYQA-UHFFFAOYSA-M
Synonym 2,6-dichloroindophenol sodium salt dihydrate, 2,6-dichloro-4-4-hydroxyphenyl azamethylene cyclohexa-2,5-dien-1-one, oxamet hane, oxamethane, sodium salt, c12h6cl2no2.na.2h2o, 2,6-dichlorophenol-indophenol sodium salt dihydrate, sodium 4-3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene amino phenolate dihydrate, sodium 4-3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene amino benzenolate dihydrate
PubChem CID 23696612
IUPAC Name sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate;dihydrate
SMILES [Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1

Specifications

Assay Percent Range 90+%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in alcohol
Formula Weight 290.07
Percent Purity 90+%
Chemical Name or Material 2, 6-Dichloroindophenol, sodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.