missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenolindophenol (Technical Grade), TRC

Product Code. 30366845
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366845 100 mg 100mg
30350309 500 mg -
30351090 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366845 Supplier Toronto Research Chemicals Supplier No. TRCC177310100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 956-48-9
Molecular Formula C12H7Cl2NO2
Molecular Weight (g/mol) 268.1
Synonym 2,5-Cyclohexadien-1-one, 2,6-dichloro-4-[(4-hydroxyphenyl)imino]-, ndophenol, 2,6-dichloro- (8CI), 2,6-Dichloro-4-[(4-hydroxyphenyl)imino]-2,5-cyclohexadien-1-one, 2,6-Dichloroindophenol, DCIP, DCPIP, DPIP, Dichloroindophenol, Dichlorophenolindophenol, Dichlorophenylindophenol, Indochlorophenol, Tillmans Reagent, p-Hydroxyphenyl-2,6-dichloro-p-benzoquinoneimine
IUPAC Name 2,6-dichloro-4-(4-hydroxyphenyl)iminocyclohexa-2,5-dien-1-one
SMILES Oc1ccc(cc1)N=C2C=C(Cl)C(=O)C(=C2)Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H7Cl2NO2/c13-10-5-8(6-11(14)12(10)17)15-7-1-3-9(16)4-2-7/h1-6,16H
Formula Weight 266.9854
Chemical Name or Material 2,6-Dichlorophenolindophenol (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.