missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenyl isocyanate, 98%

Product Code. 11338425
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338425 1 g 1g
11348425 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338425 Supplier Thermo Scientific Alfa Aesar Supplier No. L10421.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39920-37-1 | C7H3Cl2NO | 188.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39920-37-1
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.007
MDL Number MFCD00002003
InChI Key HMVKMAMIRAVXAN-UHFFFAOYSA-N
Synonym 2,6-dichlorophenyl isocyanate, benzene, 1,3-dichloro-2-isocyanato, 2,6-dichlorophenylisocyanate, isocyanic acid 2,6-dichlorophenyl ester, pubchem23916, acmc-1ailv, 2,6 dichlorophenylisocyanate, 2,6-dichlorobenzenisocyanate, 2,6dichlorophenyl isocyanate, 1,3-dichloro-2-isocyanato-benzene
PubChem CID 604537
IUPAC Name 1,3-dichloro-2-isocyanatobenzene
SMILES C1=CC(=C(C(=C1)Cl)N=C=O)Cl

Specifications

Melting Point 42°C to 46°C
Boiling Point 101°C (5 mmHg)
Flash Point 76°C (168°F)
Quantity 1 g
UN Number UN2250
Beilstein 608323
Formula Weight 188.01
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorophenyl isocyanate

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.