missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylacetic acid, 98%

Product Code. 11499287
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499287 25 g 25g
11424060 5 g 5g
11409297 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499287 Supplier Thermo Scientific Alfa Aesar Supplier No. B21354.14
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6575-24-2 | C8H6Cl2O2 | 205.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6575-24-2
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00004320
InChI Key SFAILOOQFZNOAU-UHFFFAOYSA-N
Synonym 2,6-dichlorophenylacetic acid, 2-2,6-dichlorophenyl acetic acid, 2,6-dichlorophenyl acetic acid, benzeneacetic acid, 2,6-dichloro, 2,6-dichloro benzeneacetic acid, 2,6-dichlorophenylaceticacid, unii-k8g025cx8w, 2,6-dichlorobenzeneaceticacid, 2cl
PubChem CID 81058
IUPAC Name 2-(2,6-dichlorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1)Cl)CC(=O)O)Cl

Specifications

Melting Point 158°C to 161°C
Quantity 25 g
Beilstein 1952744
Formula Weight 205.04
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorophenylacetic acid
compliance-icons
Product Identifier
  • 2,6-Dichlorophenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2,6-Dichlorophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.