missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylacetic Acid Methyl Ester, TRC

Product Code. 30385453
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30385453

Brand: Toronto Research Chemicals TRCD435655250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,6-Dichlorophenylacetic Acid Methyl Ester
CAS 54551-83-6
Molecular Formula C9 H8 Cl2 O2
Quantity 250 mg
Synonym 2,6-Dichloro-benzeneacetic Acid Methyl Ester, Methyl (2,6-Dichlorophenyl)acetate
InChI Formula InChI=1S/C9H8Cl2O2/c1-13-9(12)5-6-7(10)3-2-4-8(6)11/h2-4H,5H2,1H3
SMILES COC(=O)Cc1c(Cl)cccc1Cl
IUPAC Name methyl 2-(2,6-dichlorophenyl)acetate
Molecular Weight (g/mol) 219.06
Formula Weight 217.9901
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.