missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichlorophenylacetic Acid Methyl Ester, TRC
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
Specifications
Specifications
| Chemical Name or Material | 2,6-Dichlorophenylacetic Acid Methyl Ester |
| CAS | 54551-83-6 |
| Molecular Formula | C9 H8 Cl2 O2 |
| Quantity | 10 g |
| Synonym | 2,6-Dichloro-benzeneacetic Acid Methyl Ester, Methyl (2,6-Dichlorophenyl)acetate |
| InChI Formula | InChI=1S/C9H8Cl2O2/c1-13-9(12)5-6-7(10)3-2-4-8(6)11/h2-4H,5H2,1H3 |
| SMILES | COC(=O)Cc1c(Cl)cccc1Cl |
| IUPAC Name | methyl 2-(2,6-dichlorophenyl)acetate |
| Molecular Weight (g/mol) | 219.06 |
| Formula Weight | 217.9901 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction