missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropyrazine, 99+%

Product Code. 10772081 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10772081 1 g Glass bottle 1g
10348320 5 g Glass bottle 5g
10056193 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10772081 Supplier Thermo Scientific Acros Supplier No. 232540010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4774-14-5 | C4H2Cl2N2 | 148.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4774-14-5
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.98
InChI Key LSEAAPGIZCDEEH-UHFFFAOYSA-N
Synonym pyrazine, 2,6-dichloro, 2,6-chloro pyrazine, 2,6-dichloro pyrazine, 2,6-dichloro-pyrazine, 2-chloro-6-chloro-pyrazine, 2,6-dicloropyrazine, 2,6 dichloropyrazine, pubchem8548, 2,6-dichloro-pyrazin, 2, 6-dichloropyrazine
PubChem CID 78504
IUPAC Name 2,6-dichloropyrazine
SMILES C1=C(N=C(C=N1)Cl)Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 53.0°C to 56.0°C
Color White to Beige
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 23, III, 903
Formula Weight 148.98
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2, 6-Dichloropyrazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.