missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichlorotoluene-3,4,5-d3, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | 2,6-Dichlorotoluene-3,4,5-D3 |
| CAS | 358731-95-0 |
| Molecular Formula | C7 D3 H3 Cl2 |
| Quantity | 1 g |
| Synonym | Benzene-1,2,3-d3, 4,6-dichloro-5-methyl- (9 CI), 4,6-Dichloro-5-methylbenzene-1,2,3-d3, 2,6-Dichlorotoluene-3,4,5-d3, 2,6-Dichlorotoluene-3,4,5-D3 |
| InChI Formula | InChI=1 S/C7H6Cl2/c1-5-6(8)3-2-4-7(5)9/h2-4 H,1H3/i2D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(Cl)c(C)c(Cl)c1[2 H] |
| IUPAC Name | 1,5-dichloro-2,3,4-trideuterio-6-methylbenzene |
| Molecular Weight (g/mol) | 164.047 |
| Formula Weight | 163.0035 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction