missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diethylphenyl isocyanate, 98+%

Product Code. 11367716
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367716 1 g 1g
11377716 5 g 5g
11387716 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11367716 Supplier Thermo Scientific Alfa Aesar Supplier No. L11339.03
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 20458-99-5 | C11H13NO | 175.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20458-99-5
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.23
MDL Number MFCD00019912
InChI Key AUSRFKVPUCQCCN-UHFFFAOYSA-N
Synonym 2,6-diethylphenyl isocyanate, 2,6-diethylphenylisocyanate, 1,3-diethyl-2-isocyanato-benzene, benzene, 1,3-diethyl-2-isocyanato, acmc-1co7s, 2,6-diethylbenzenisocyanate, benzene,1,3-diethyl-2-isocyanato
PubChem CID 5153183
IUPAC Name 1,3-diethyl-2-isocyanatobenzene
SMILES CCC1=CC=CC(CC)=C1N=C=O

Specifications

Density 1.014
Boiling Point 114°C to 115°C (1 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.526
Quantity 1 g
UN Number UN2206
Beilstein 2641462
Sensitivity Moisture sensitive
Formula Weight 175.23
Percent Purity ≥98%
Chemical Name or Material 2,6-Diethylphenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.