missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dimethoxyacetophenone, 98%, Thermo Scientific™

Product Code. 10502318 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
10502318 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 10502318

Brand: Thermo Scientific Acros 295130050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2040-04-2
CAS Min % 97.5
CAS Max % 100.0
Melting Point 69.5°C to 71.0°C
Color White to Beige
Boiling Point 135.0°C to 136.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Molecular Formula C10H12O3
Linear Formula (CH3O)2C6H3COCH3
MDL Number MFCD00008729
Quantity 5 g
Synonym 2',6'-dimethoxyacetophenone, 1-2,6-dimethoxyphenyl ethanone, 2,6-dimethoxyacetophenone, acetophenone, 2',6'-dimethoxy, 1-2,6-dimethoxyphenyl ethan-1-one, ethanone, 1-2,6-dimethoxyphenyl, usaf k-2801, 2,6-dimethoxy acetophenone, 1-acetyl-2,6-dimethoxybenzene, pubchem3380
InChI Key XEUGKOFTNAYMMX-UHFFFAOYSA-N
SMILES CC(=O)C1=C(C=CC=C1OC)OC
IUPAC Name 1-(2,6-dimethoxyphenyl)ethanone
Molecular Weight (g/mol) 180.21
PubChem CID 16267
Formula Weight 180.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2', 6'-Dimethoxyacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2',6'-Dimethoxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.