missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-heptanone, >90% (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone)

Product Code. 11342857
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342857 2500 mL 2.5 L
11322857 100 mL 100mL
11332857 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11342857 Supplier Thermo Scientific Alfa Aesar Supplier No. L16015.0F
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108-83-8 | C9H18O | 142.242 g/mol

2,6-Dimethyl-4-heptanone is used as a coating solvent. It is an active component of mint oil. It acts as a dispersant for organosol type resins. It is involved in the antigerminative treatment of bulbs and tubers. Further, it is used as a solvent for nitrocellulose. In addition to this, it acts as an intermediate in the preparation of inhibitors, active pharmaceutical ingredients and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethyl-4-heptanone is used as a coating solvent. It is an active component of mint oil. It acts as a dispersant for organosol type resins. It is involved in the antigerminative treatment of bulbs and tubers. Further, it is used as a solvent for nitrocellulose. In addition to this, it acts as an intermediate in the preparation of inhibitors, active pharmaceutical ingredients and dyes.

Solubility
Miscible with ethanol, ether, carbon tetrachloride, chloroform, benzene and most organic liquids.

Notes
Incompatible with strong bases, strong oxidizing agents, strong acids and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-83-8
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00008940
InChI Key PTTPXKJBFFKCEK-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4-heptanone, diisobutyl ketone, isovalerone, isobutyl ketone, valerone, diisobutylketone, diisobutilchetone, s-diisopropylacetone, di-isobutylcetone, dibk
PubChem CID 7958
IUPAC Name 2,6-dimethylheptan-4-one
SMILES CC(C)CC(=O)CC(C)C

Specifications

Melting Point -42°C
Density 0.809
Boiling Point 163°C to 173°C
Flash Point 47°C (117°F)
Odor Unpleasant
Refractive Index 1.414
Quantity 2500 mL
UN Number UN1224
Beilstein 1743163
Solubility Information Miscible with ethanol,ether,carbon tetrachloride,chloroform,benzene and most organic liquids.
Formula Weight 142.24
Percent Purity >90%
Chemical Name or Material 2,6-Dimethyl-4-heptanone, (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,6-Dimethyl-4-heptanone, (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone)
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: Diisobutyl ketone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.