missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-pyrone, 99%

Product Code. 11405163
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405163 50 g 50g
11495153 10 g 10g
11415163 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405163 Supplier Thermo Scientific Alfa Aesar Supplier No. A12799.18
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1004-36-0 | C7H8O2 | 124.139 g/mol

2,6-Dimethyl-4-pyrone is used as a chemical and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethyl-4-pyrone is used as a chemical and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressure.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1004-36-0
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00006579
InChI Key VSYFZULSKMFUJJ-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4h-pyran-4-one, 2,6-dimethyl-4-pyrone, 2,6-dimethyl-gamma-pyrone, 2,6-dimethyl-4-pyranone, 4h-pyran-4-one, 2,6-dimethyl, gamma-2,6-dimethylpyrone, unii-v2d262mf0l, 2,6-dimethyl-pyran-4-one, 2,6-dimethyl-.gamma.-pyrone, ccris 3601
PubChem CID 13862
IUPAC Name 2,6-dimethylpyran-4-one
SMILES CC1=CC(=O)C=C(O1)C

Specifications

Melting Point 132°C to 136°C
Boiling Point 248°C to 250°C
Quantity 50 g
Beilstein 107418
Solubility Information Slightly soluble in water.
Formula Weight 124.14
Percent Purity 99%
Chemical Name or Material 2,6-Dimethyl-4-pyrone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.