missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethylaniline-d11, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2,6-Dimethylaniline-d11 |
| CAS | 1092805-08-7 |
| Molecular Formula | C8 D11 N |
| Quantity | 0.1 g |
| Synonym | Benzen-3,4,5-d3-amine-d2, 2,6-di(methyl-d3)- (ACI), 2,6-Di(methyl-d3)benzen-3,4,5-d3-amine-d2 (ACI), 2,6-Dimethylaniline-d11, 2,6-Dimethylaniline-D11 |
| InChI Formula | InChI=1 S/C8H11N/c1-6-4-3-5-7(2)8(6)9/h3-5 H,9H2,1-2H3/i1D3,2D3,3 D,4 D,5 D/hD2 |
| SMILES | [2 H]N([2 H])c1c(c([2 H])c([2 H])c([2 H])c1C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | N,N,3,4,5-pentadeuterio-2,6-bis(trideuteriomethyl)aniline |
| Molecular Weight (g/mol) | 132.2474 |
| Formula Weight | 132.1582 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction