missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylanisole, 98+%

Product Code. 11464267
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464267 25 g 25g
11474267 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464267 Supplier Thermo Scientific Alfa Aesar Supplier No. B20365.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1004-66-6 | C9H12O | 136.19 g/mol

2,6-Dimethylanisole is used as a pharmaceutical intermediate. Methoxymetacyclophanes is prepared from 2,6-dimethylanisole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethylanisole is used as a pharmaceutical intermediate. Methoxymetacyclophanes is prepared from 2,6-dimethylanisole.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1004-66-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00008380
InChI Key GFNZJAUVJCGWLW-UHFFFAOYSA-N
Synonym 2,6-dimethylanisole, 2-methoxy-m-xylene, benzene, 2-methoxy-1,3-dimethyl, 1,3-dimethyl-2-methoxybenzene, pubchem4115, 2,6-dimethyl anisole, acmc-1buuc, ksc493q8r, 2,6-dimethylphenyl methyl ether
PubChem CID 66088
IUPAC Name 2-methoxy-1,3-dimethylbenzene
SMILES COC1=C(C)C=CC=C1C

Specifications

Density 0.962
Boiling Point 181°C to 182°C
Quantity 25 g
Refractive Index 1.503
Beilstein 1858509
Solubility Information Insoluble in water.
Formula Weight 136.19
Percent Purity ≥98%
Chemical Name or Material 2,6-Dimethylanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.