missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylcyclohexanol, 99%, mixture of isomers

Product Code. 10673363 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
This item is not returnable. View return policy

Product Code. 10673363

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5337-72-4 | C8H16O | 128.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5337-72-4
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.21
MDL Number MFCD00001502
InChI Key MOISVRZIQDQVPF-UHFFFAOYSA-N
Synonym 2,6-dimethylcyclohexanol, cyclohexanol, 2,6-dimethyl, 2,6-dimethylcyclohexyl alcohol, ambsclk-255, cyclohexanol,6-dimethyl, 2,6-dimethyl-cyclohexanol, acmc-20ao81, 2,6-dimethylcyclohexanol,c&t, 2,6-dimethyl-cyclohexan-1-ol, 2,6-dimethylcyclohexanol, mixture of isomers
PubChem CID 21428
IUPAC Name 2,6-dimethylcyclohexan-1-ol
SMILES CC1CCCC(C1O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9400g/mL
Boiling Point 174.4°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2C6H9OH
Refractive Index 1.4585 to 1.4615
Quantity 5 g
Beilstein 06, 18
Specific Gravity 0.94
Formula Weight 128.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 6-Dimethylcyclohexanol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dimethylcyclohexanol, mixture of isomers
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.