missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethylcyclohexanol, TRC
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | 2,6-Dimethylcyclohexanol |
| CAS | 5337-72-4 |
| Molecular Formula | C8 H16 O |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C8H16O/c1-6-4-3-5-7(2)8(6)9/h6-9H,3-5H2,1-2H3 |
| SMILES | CC1CCCC(C)C1O |
| IUPAC Name | 2,6-dimethylcyclohexan-1-ol |
| Molecular Weight (g/mol) | 128.212 |
| Formula Weight | 128.12 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction