missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenyl isocyanide

Product Code. 11347967
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11347967

Brand: Thermo Scientific Alfa Aesar 020748.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2769-71-3 | C9H9N | 131.18 g/mol

2,6-Dimethylphenyl isocyanate is used in the preparation of homoleptic metallaamidine complex, derivatized bets-cyclodextrins. It is used as chiral stationary phase in normal-phase liquid chromatography. It is also involved in the preparation of tris( 2,6-dimethylphenylimido)methylrhenium (VII) and 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2769-71-3
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00013479
InChI Key DNJLFZHMJDSJFN-UHFFFAOYSA-N
Synonym 2,6-dimethylphenyl isocyanide, 2,6-dimethylphenylisonitrile, 2,6-dimethylphenylisocyanide, 2,6-xyleneisonitrile, 2,6-xylyl isocyanide, benzene,2-isocyano-1,3-dimethyl, benzene, 2-isocyano-1,3-dimethyl-9ci, vic.-m-xylyl isocyanide, acmc-20ao80, 2,6-dimethyiphenyl isocyanide
PubChem CID 76009
IUPAC Name 2-isocyano-1,3-dimethylbenzene
SMILES CC1=CC=CC(C)=C1[N+]#[C-]

Specifications

Melting Point 73°C to 74°C
Quantity 5 g
Odor Unpleasant
Physical Form Crystalline
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 131.18
Chemical Name or Material 2,6-Dimethylphenyl isocyanide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.