missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10291323
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10291323 10 g 10g
10002514 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10291323 Supplier Thermo Scientific Maybridge Supplier No. SB01642EA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 19241-16-8
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.238
InChI Key UULUECCNPPJFBU-UHFFFAOYSA-N
Synonym 2,6-dimethylphenyl isothiocyanate, 2,6-xylyl isothiocyanate, 2,6-dimethylphenylisothiocyanate, benzene, 2-isothiocyanato-1,3-dimethyl, 2-isothiocyanato-1,3-dimethyl-benzene, benzene,2-isothiocyanato-1,3-dimethyl, 2,6-dimethylbenzenisothiocyanate, acmc-20ao7z, 2,6-dimethylphenyl-isothiocyanate, 2,6-dimethylphenyl-1-isothiocyanate
PubChem CID 87976
IUPAC Name 2-isothiocyanato-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)N=C=S

Specifications

Quantity 10 g
Assay Percent Range 97%
Formula Weight 163.24
Chemical Name or Material 2, 6-Dimethylphenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.