missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenyl isothiocyanate, 98%

Product Code. 11424057
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424057 5 g 5g
11434057 25 g 25g
11444057 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424057 Supplier Thermo Scientific Alfa Aesar Supplier No. A19480.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 19241-16-8 | C9H9NS | 163.238 g/mol

2,6-Dimethylphenyl isocyanate has been used in the preparation of derivatized β-cyclodextrins, used as chiral stationary phase in normal-phase liquid chromatography, tris( 2,6-dimethylphenylimido)methylrhenium (VII), 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethylphenyl isocyanate has been used in the preparation of derivatized β-cyclodextrins, used as chiral stationary phase in normal-phase liquid chromatography, tris( 2,6-dimethylphenylimido)methylrhenium (VII), 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19241-16-8
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.238
MDL Number MFCD00038715
InChI Key UULUECCNPPJFBU-UHFFFAOYSA-N
PubChem CID 87976
IUPAC Name 2-isothiocyanato-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)N=C=S

Specifications

Density 1.085
Quantity 5 g
Odor Pungent
Refractive Index 1.625
UN Number UN2922
Beilstein 637195
Synonym 2,6-dimethylphenyl isothiocyanate, 2,6-xylyl isothiocyanate, 2,6-dimethylphenylisothiocyanate, benzene, 2-isothiocyanato-1,3-dimethyl, 2-isothiocyanato-1,3-dimethyl-benzene, benzene,2-isothiocyanato-1,3-dimethyl, 2,6-dimethylbenzenisothiocyanate, acmc-20ao7z, 2,6-dimethylphenyl-isothiocyanate, 2,6-dimethylphenyl-1-isothiocyanate
Boiling Point 247°C
Solubility Information Hydrolyzes in water.
Formula Weight 163.24
Percent Purity 98%
Chemical Name or Material 2,6-Dimethylphenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.