missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenylmagnesium bromide, 1M solution in THF, AcroSeal™, Thermo Scientific™

Product Code. 11935981 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
100mL; AcroSeal glass bottle
This item is not returnable. View return policy

Product Code. 11935981

Brand: Thermo Scientific Acros 436061000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 21450-64-6, 109-99-9
CAS Min % 19.93, 76.92
CAS Max % 23.08, 80.07
Quantity 100mL
Molecular Formula C8H9BrMg
Flash Point −19°C
Synonym 2,6-dimethylphenylmagnesium bromide, 2,6-dimethylphenylmagnesium bromide solution, biguvduzgajwqk-uhfffaoysa-m, 2,6-dimethylphenymagnesium bromide, bromo 2,6-dimethylphenyl magnesium, 2,6-dimethyl-phenylmagnesium bromide, 2,6-dimethylphenyl magnesium bromide, 2,6-dimethylphenylmagnesium bromide, 0.5m in 2-methf, 2,6-dimethylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 2,6-dimethylphenylmagnesium bromide solution, 1.0 m in thf, grignard reagent
Solubility Information Solubility in water: reacts.
InChI Key DCQJQVFIEFMTTQ-UHFFFAOYSA-M
SMILES CC1=[C-]C(=CC=C1)C.[Mg+2].[Br-]
IUPAC Name magnesium;1,3-dimethylbenzene-2-ide;bromide
Molecular Weight (g/mol) 209.369
PubChem CID 4517616
Formula Weight 209.37
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.998
Density 0.9980g/mL
Chemical Name or Material 2,6-Dimethylphenylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,6-Dimethylphenylmagnesium bromide, 1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.