missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dinitrotoluene, 97%

Product Code. 11354324
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354324 5 g 5g
11364324 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354324 Supplier Thermo Scientific Alfa Aesar Supplier No. L00856.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 606-20-2 | C7H6N2O4 | 182.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-20-2
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.135
MDL Number MFCD00007158
InChI Key XTRDKALNCIHHNI-UHFFFAOYSA-N
Synonym 2,6-dinitrotoluene, 2,6-dnt, benzene, 2-methyl-1,3-dinitro, 1-methyl-2,6-dinitrobenzene, 2,6-dinitromethylbenzene, toluene, 2,6-dinitro, rcra waste number u106, 2-methyl-1,3-dinitro-benzene, unii-gg7fav92mk, ccris 1006
PubChem CID 11813
ChEBI CHEBI:957
IUPAC Name 2-methyl-1,3-dinitrobenzene
SMILES CC1=C(C=CC=C1[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 58°C to 65°C
Boiling Point 285°C
Flash Point 207°C (404°F)
Quantity 5 g
UN Number UN3454
Beilstein 2052046
Formula Weight 182.14
Percent Purity 97%
Chemical Name or Material 2,6-Dinitrotoluene
compliance-icons compliance-icons
Product Identifier
  • 2,6-Dinitrotoluene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: 2,6-Dinitrotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.