missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diphenylphenol, TRC

Product Code. 30378979
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378979 1 g 1g
30377629 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30378979 Supplier Toronto Research Chemicals Supplier No. TRCD4922301G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2432-11-3
Molecular Formula C18H14O
Molecular Weight (g/mol) 246.3
Synonym [1,1':3',1''-Terphenyl]-2'-ol, [m-Terphenyl]-2'-ol, 2'-Hydroxy-m-terphenyl
SMILES Oc1c(cccc1c2ccccc2)c3ccccc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H14O/c19-18-16(14-8-3-1-4-9-14)12-7-13-17(18)15-10-5-2-6-11-15/h1-13,19H
Formula Weight 246.1045
Chemical Name or Material 2,6-Diphenylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.