missing translation for 'onlineSavingsMsg'
Learn More

2-(6-Methoxynaphthalen-2-yl)propanenitrile, TRC

Product Code. 30471455
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471455 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471455 Supplier Toronto Research Chemicals Supplier No. TRCM266735500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(6-Methoxynaphthalen-2-yl)propanenitrile
CAS 86603-94-3
Molecular Formula C14 H13 N O
Quantity 500 mg
Synonym 2-Naphthaleneacetonitrile','6-methoxy-α-methyl- (9CI','ACI), 6-Methoxy-α-methyl-2-naphthaleneacetonitrile (ACI), (±)-Naproxen nitrile, 2-(6-Methoxy-2-naphthyl)propionitrile, α-Methyl-6-methoxy-2-Naphthaleneacetonitrile
InChI Formula InChI=1S/C14H13NO/c1-10(9-15)11-3-4-13-8-14(16-2)6-5-12(13)7-11/h3-8,10H,1-2H3
SMILES COc1ccc2cc(ccc2c1)C(C)C#N
IUPAC Name 2-(6-methoxy-2-naphthyl)propanenitrile
Molecular Weight (g/mol) 211.26
Formula Weight 211.0997
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.