missing translation for 'onlineSavingsMsg'
Learn More

2',7'-Dichlorofluorescein diacetate, TRC

Product Code. 30136155
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136155 50 mg 50mg
30134089 1 g 1g
30009769 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136155 Supplier Toronto Research Chemicals Supplier No. TRCD43453350MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2044-85-1
Molecular Formula C24H14Cl2O7
Molecular Weight (g/mol) 485.27
Synonym MFCD 37501, 3',6'-Bis(acetyloxy)-2',7'-dichlorospiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one
SMILES CC(=O)Oc1cc2Oc3cc(OC(=O)C)c(Cl)cc3C4(OC(=O)c5ccccc45)c2cc1Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H14Cl2O7/c1-11(27)30-21-9-19-15(7-17(21)25)24(14-6-4-3-5-13(14)23(29)33-24)16-8-18(26)22(31-12(2)28)10-20(16)32-19/h3-10H,1-2H3
Formula Weight 484.0117
Chemical Name or Material 2',7'-Dichlorofluorescein diacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.