missing translation for 'onlineSavingsMsg'
Learn More

2',7'-Dichlorofluorescein

Product Code. 11403307
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11403307 25 g 25g
11493297 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11403307 Supplier Thermo Scientific Alfa Aesar Supplier No. A16212.14
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 76-54-0 | C20H10Cl2O5 | 401.195 g/mol

2',7'-Dichlorofluorescein is used as an indicator and as a cellular antioxidant activity. It is also used for the determination of carbohydrates and hydrogen peroxide by fluorescence densitometry and chemiluminescence respectively. It is used as an indicator for argentometry by Fajans method. It is also involved in the cellular antioxidant activity assay. It is utilized as an additive in fluorescence and laser studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,7′-Dichlorofluorescein is used as an indicator and as a cellular antioxidant activity. It is also used for the determination of carbohydrates and hydrogen peroxide by fluorescence densitometry and chemiluminescence respectively. It is used as an indicator for argentometry by Fajans method. It is also involved in the cellular antioxidant activity assay. It is utilized as an additive in fluorescence and laser studies.

Solubility
Soluble in ethanol and methanol. Slightly dimethyl sulfoxide. Insoluble in water.

Notes
As a fluorescent indicatorIncompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-54-0
Molecular Formula C20H10Cl2O5
Molecular Weight (g/mol) 401.195
MDL Number MFCD00005047
InChI Key VFNKZQNIXUFLBC-UHFFFAOYSA-N
Synonym 2',7'-dichlorofluorescein, dichlorofluorescein, unii-56nqm5uzt1, 2,7-dichlorofluorescein, 56nqm5uzt1, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 2',7'-dichloro-3',6'-dihydroxy, 2',7'-dichloro-3',6'-dihydroxy-3h-spiro isobenzofuran-1,9'-xanthen-3-one, 2',7'-dichloro-3',6'-dihydroxyspiro 2-benzofuran-3,9'-xanthene-1-one, 11,16-dichloro-12,15-dihydroxyspiro 3-hydroisobenzofuran-3,9'-xanthene-1-one
PubChem CID 64944
ChEBI CHEBI:51596
IUPAC Name 2',7'-dichloro-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)Cl)O)O)Cl

Specifications

Melting Point >300°C (decomposition)
Quantity 25 g
Solubility Information Soluble in ethanol and methanol. Slightly dimethyl sulfoxide. Insoluble in water.
Formula Weight 401.2
Chemical Name or Material 2',7'-Dichlorofluorescein

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.